Learn More
Masterflex™ Nylon Barbed Elbow Connector
Create ninety-degree bends in flexible tubing without the kinks
Brand:  Masterflex™ 30622-81
Product Code. 15394178
  • 30.00€ / Pack of 10
Only null left
Estimated Shipment: 19-11-2025
Description
  • 90° bend tubing connector
  • Barb connector
Specifications
Specifications
90° elbow tubing connector
Nylon
2-port, 1/2 in. I.D. tubing
Not Autoclavable
Flexible tubing
EtO only