Learn More
Thermo Scientific™ Nunc™ PETG Roller Bottles
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.
187.00€ - 1000.00€
Specifications
Color | Clear |
---|---|
Material | PETG |
Closure Included | Yes |
Graduated | Yes |
Shape | Round |
Product Code | Brand | Cell Growth Area | Bottle Surface | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cell Growth Area | Bottle Surface | Packaging | Price | Quantity & Availability | |||
10059702
|
Thermo Scientific™
1060-05 |
1050 cm2 | Smooth | 5/sleeve, 20/case |
187.00€
Pack of 20 |
In Stock
Log in to see stock. |
|||
10712974
|
Thermo Scientific™
1060-20 |
1050 cm2 | Smooth | 20/case |
210.00€
Pack of 20 |
In Stock
Log in to see stock. |
|||
10263393
|
Thermo Scientific™
1060-85 |
1050 cm2 | Smooth | 5/sleeve, 20/case |
285.00€
Pack of 20 |
Estimated Shipment: 02-10-2025
Log in to see stock. |
|||
10627584
|
Thermo Scientific™
1760-20 |
1700 cm2 | Pleated | 20/case |
395.00€
Pack of 20 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||
10385073
|
Thermo Scientific™
1860-22 |
1800 cm2 | Smooth | 22/case |
518.00€
Pack of 22 |
In Stock
Log in to see stock. |
|||
10704144
|
Thermo Scientific™
2160-20 |
2100 cm2 | Pleated | 20/case |
518.00€
Pack of 20 |
Estimated Shipment: 14-10-2025
Log in to see stock. |
|||
10551844
|
Thermo Scientific™
2160-05 |
2100 cm2 | Pleated | 5/sleeve, 20/case |
518.00€
Pack of 20 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||
10596895
|
Thermo Scientific™
4260-22 |
4200 cm2 | Pleated | 22/case |
1000.00€
Pack of 22 |
Estimated Shipment: 06-11-2025
Log in to see stock. |
|||
Description
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor with Thermo Scientific™ Nunc™ PETG Roller Bottles. Virtually unbreakable PETG provides added assurance that valuable contents are safe during laboratory and industrial-scale applications including vaccines, cell culture expansion and production of other biologics.
Key Features:- Molded of durable PETG
- Can be frozen down to -40°C — supports freeze thaw cell release methods, reducing reliance on trypsin
- Provides solutions for all scales with wide range of surface areas from 1050cm2 to 4200cm2
- Reduces wrist strain and increases productivity with quick-action ergonomic closure
- Can be traced by lot number on every bottle
- Cell culture expansion
- Vaccine production
- Antibody production
Specifications
Clear | |
Yes | |
Round | |
Quick-Action Ergonomic |
PETG | |
Yes | |
Roller Bottle |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.