Learn More
Scientific Industries SI™ Multiple Sample Starter Set
Holds samples in place while mixing. Scientific Industries SI™ Multiple Sample Starter Set includes one microtube foam insert and one microplate foam insert that will accommodate either 60 microtubes or one standard footprint microplate.
Brand:  Scientific Industries SI™ 504-0233-00
Product Code. 10223121
  • 105.00€ / Each
Only null left
Estimated Shipment: 29-09-2025
Description
For use with Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse
  • 152mm Platform
  • Microtube Foam Insert
  • Microplate Foam Insert
Specifications
Specifications
Vortex-Genie 2 family of mixers and Vortex-Genie Pulse
  • 1 Microtube Foam Insert - accommodates 60 microtubes
  • 1 Microplate Foam Insert - accommodates 1 standard footprint microplate
Platform Attachment 15577315