Learn More
Eppendorf™ Multi-Position Wire Shelf Rack
For Use With: Innova CO-14 Incubator
Brand:  Eppendorf™ P0628-6170
399.96 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11340045
  • 421.68 EUR / Each
Only null left
In Stock
Description
  • Multi-Position Wire Shelf Rack, 4 positions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Innova CO-14 Incubator
Shelf Rack