Learn More
MP Biomedicals™ Microplate Sealing Tape
Shop All MP Biomedical Products
Click to view available options
Quantity:
1 Roll
Unit Size:
1 roll
Specifications
Specifications
Material | Acetate |
No. per Case | Each |
Quantity | 1 Roll |
For Use With (Application) | Suitable for all microplates for both short and long term storage, providing a tight seal to minimize evaporation and condensation |
Product Type | Microplate Sealing Tape |