Learn More
MP Biomedicals™ 1X Hanks' Balanced Salt Solution (Modified) without Phenol Red
Shop All MP Biomedical Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- Phenol red has been shown to interfere with the growth of some cells at cloning densities.
- Use this medium when working with stem cells or when growing cells at low densities.
Specifications
Specifications
Chemical Name or Material | Hanks' Balanced Salt Solution (HBSS) |
Color | Clear |
Concentration | 1 X |
pH | 6.9 to 7.4 |
Physical Form | Liquid |
Quantity | 500 mL |
Formulation | Calcium Chloride 140 mg/L, Magnesium Sulfate 97.7 mg/L, Potassium Chloride 400 mg/L, Potassium Phosphate Monobasic 60 mg/L, Sodium Bicarbonate 350 mg/L, Sodium Chloride 8000 mg/L, Sodium Phosphate Dibasic 47.7 mg/L, Dextrose 1000 mg/L |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.