Learn More
DuPont™ Tyvek™ 500 Xpert Hooded Coverall Green
The Tyvek™ 500 Xpert Hooded Coverall is composed of flash-spun HDPE, providing an ideal balance of protection, durability and comfort. The 3-piece hood provides optimal fit to head and face when turning while ample crotch area offers freedom of movement.
1005.00€
Specifications
Product Type | Hooded Coveralls |
---|---|
Material | HDPE |
Color | Green |
Waist Style | Elastic |
Ankle Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
13436646
![]() |
DuPont™
D14936675 |
X-Large |
1005.00€
Pack of 100 |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
13416646
![]() |
DuPont™
D14936650 |
Medium |
1005.00€
Pack of 100 |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
13456646
![]() |
DuPont™
D14936681 |
2X-Large |
1005.00€
Pack of 100 |
Estimated Shipment: 02-12-2025
Log in to see stock. |
|||||
13446646
![]() |
DuPont™
D14936664 |
Large |
1005.00€
Pack of 100 |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
Description
- Coverall with 3-piece hood for optimal fit to head and face when turning.
- Excellent barrier against fine particles and fibers down to 1 micron in size.
- Ergonomic-protective design.
- Stitched external seams.
- Elasticated face, wrists and ankles as well as glued-in waist elastic.
- Large, easy-to-grasp zipper puller.
- Robust yet lightweight (<180 g per garment).
- Ample crotch area for freedom of movement.
- Ultra-low-linting and antistatically treated.
Specifications
Hooded Coveralls | |
Green | |
Elastic | |
Hooded | |
Zipper | |
Category III, Type 5 and 6 | |
(EU) 2016/425, EN 14126, EN 1073-2 |
HDPE | |
Elastic | |
Elastic | |
3-panel | |
Stitched | |
Abrasion |