Learn More
Thermo Scientific™ Mini Capillary Unions
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
71.65€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10311155
|
Thermo Scientific™
290GU498 |
0.22 to 0.32 mm ID Capillary GC Column |
71.65€
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||||
10289074
|
Thermo Scientific™
290GU499 |
0.53 mm ID Capillary GC Column |
71.65€
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
Description
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Kit contains one union and five ferrulesIncludes:
Union, 1; Ferrules, 5
Specifications
Capillary Union |