Learn More
Hirschmann™ MicroPipetter
Click to view available options
For Use With (Application):
Micropipettes
Unit Size:
Each
Description
- Contains replaceable adapter for optimum adaptation to pipette size
Specifications
Specifications
For Use With (Application) | Micropipettes |
Includes | Ring Mark and replaceable adapter |
Type | Micropipet |
Disposable | Single-use |
For Use With (Equipment) | Micropipettes |