Learn More
Ansell™ Microchem™ 4000 PAPR Encapsulated Coveralls
MICROCHEM™ 4000 PAPR Encapsulated Coveralls provide complete protection from liquid and particulate hazards. Ventilated suits with filtered air and MICROGARD™/MICROCHEM™ technology provide head and body protection from hazardous substances.
759.00€
Specifications
Certifications/Compliance | Type 3/4 EN14605, Type 5 EN ISO 13982-1, Types 3-B, 4-B EN14126, EN1073-2, EN1149-5 |
---|---|
Color | Green |
Hood Type | Fully Encapsulated |
Material | Multi-layer Barrier Laminate |
Ankle Style | Attached socks, Boot overflaps |
Description
- Fully Encapsulated Coverall, for use in combination with Scott Proflow 2SC/EX PAPR fan unit, breathing hose & filters.
- Double elasticated cuffs, attached socks with boot overflap.
- Dual zip front entry
- Emergency rip cords attached to left chest shoulder and left back shoulder as worn.
- Permeation tested against over 180 chemicals, including chemical warfare agents
- Textile-like inner provides wearer acceptance
- Anti-static according to EN 1149-5
Specifications
Type 3/4 EN14605, Type 5 EN ISO 13982-1, Types 3-B, 4-B EN14126, EN1073-2, EN1149-5 | |
Fully Encapsulated | |
Attached socks, Boot overflaps | |
Double zipper front entry | |
Emergency rip cords, attached socks with boot overflap | |
Welded, Taped |
Green | |
Multi-layer Barrier Laminate | |
Accommodates Scott Proflow 2SC/EX PAPR fan unit, breathing hose and filters | |
Double, Elastic | |
None | |
Includes Emergency rip cords, attached socks with boot overflap |