Learn More
Miele Professional™ Membrane Pump and Suction Lance
Extra disp. canisters with installation by customer service
374.00€ - 396.67€
Specifications
Product Type | Dispensing System |
---|---|
For Use With (Equipment) | Washer-Disinfectors |
Quantity | 1 Ea. |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
30533230
|
Miele Professional™
12426680 |
APLW 883 |
374.00€
1 sheet |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30533227
|
Miele Professional™
12426740 |
APLW 884 |
385.33€
1 sheet |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30533234
|
Miele Professional™
12426770 |
APLW 885 |
396.67€
1 sheet |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For 5 - 10 L canisters in the dispensing drawer
- The suction lance features level detection
- Installed by Customer Service
- Extra disp. canisters with installation by customer service
Specifications
Dispensing System | |
Washer-Disinfectors | |
1 Ea. |