Learn More
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
11907711
|
Mettler Toledo™
51107688 |
Barium |
631.00€
1 set |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
11907721
|
Mettler Toledo™
51107874 |
Lead |
631.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11917711
|
Mettler Toledo™
51107690 |
Fluoroborate |
631.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11927711
|
Mettler Toledo™
51107689 |
Sulfide |
631.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11937711
|
Mettler Toledo™
51107692 |
Copper |
641.00€
1 set |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
11947711
|
Mettler Toledo™
51107694 |
Iodide |
636.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11957711
|
Mettler Toledo™
51107695 |
Cyanide |
631.00€
1 set |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
11977711
|
Mettler Toledo™
51107871 |
Thiocyanate |
631.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11987701
|
Mettler Toledo™
51107686 |
Cadmium |
631.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11997701
|
Mettler Toledo™
51107687 |
Lithium |
631.00€
1 set |
In Stock
Log in to see stock. |
|||||
Description
Specifications
Membrane Kit |