Learn More
3M™ Maxim Series Polycarbonate Lens Safety Goggles
3.15€ - 12.60€
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
---|---|
Certifications/Compliance | EN 166: 2001 |
Product Type | Safety goggles |
Frame Color | Black |
No. per Pack | 1 Ea. |
Product Code | Brand | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Lens Tint | Price | Quantity & Availability | |||||
12632545
|
3M™
7010045682 |
Clear | N/A | ||||||
12622605
|
3M™
7010045680 |
Bronze |
3.15€
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||||
12632615
|
3M™
7010045727 |
Amber |
12.60€
Each Qty Price Break Available |
In Stock
Log in to see stock. ![]() |
|||||
Description
Specifications
Anti-fog, Scratch-resistant | |
Safety goggles | |
1 Ea. |
EN 166: 2001 | |
Black | |
Polycarbonate |