Learn More
Brady™ Magnetic Plastic ADR Transport Signs
Brand: Brady™ 223590
28.79 EUR valid until 2025-08-24
Use promo code "24132" to get your promotional price.
This item is not returnable.
View return policy
Specifications
ADR 1.4-297*297-B0859 | |
1 Ea. | |
297 mm | |
Diamond | |
Black on Orange |
Magnetic Plastic | |
Permanent Acrylic | |
297.00 mm | |
1.4 1 |