Learn More
Motic™ LWD Condenser for AE2000
Long working distance condenser suitable for phase contrast. Motic™ LWD Condenser for AE2000, when used with phase accessories, facilitates visualization of transparent specimens.
Brand: Motic™ 1101000201551
550.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Concentrates and focuses light passing through the specimen.
- Long working-distance condenser.
- Suitable for phase contrast.
Specifications
NA: 0.40 | |
LWD Condenser | |
AE2000 Microscopes | |
28 mm |