Learn More
Witeg™ Low Form Glass Beakers with Spout
With graduations for accurate and precise filling and dispensing.
Brand:  Witeg™ 5 500 250 S
 View more versions of this product
Product Code. 13417547
  • 23.10€ / Pack of 10
Only null left
Estimated Shipment: 02-10-2025
Description
  • Wide range of capacities available
  • Acc. to ISO 3819
  • Manufactured from DURAN™ borosilicate glass 3.3
  • With spout for easy pouring
Specifications
Specifications
Transparent
250 mL
95 mm
Beaker
ISO 3819
70 mm
Borosilicate Glass
Spout
Yes
Low Form