Learn More
Fisherbrand™ Leaf Calibration Weight Set, Class M1
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
Brand:  Fisherbrand™ FB67829
87.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15585109
  • 121.00€ / 1 unit
Only null left
Estimated Shipment: 22-09-2025
Add to basket
Description
Description
  • Class M1 calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
M1
1 to 500 mg
1 mg to 500 mg