Learn More
Fisherbrand™ Leaf Calibration Weight Set, Class F2 with certificate
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
Brand:  Fisherbrand™ FB67827
165.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15565109
  • 229.00€ / 1 unit
Only null left
Estimated Shipment: 05-09-2025
Add to basket
Description
Description
  • Class F2 calibration weight
  • Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
F2
1 to 500 mg
1 mg to 500 mg