Learn More
Buerkle™ Polyethylene Square All-Purpose Boxes
All-purpose boxes are used for storing, packing, taking samples, sampling, and transport of pastes, granules, powders, soil samples and frozen goods.
3.75€ - 14.20€
Specifications
Closure Type | Press-in Lid |
---|---|
Color | Transparent |
Material | Polyethylene |
Shape | Square |
For Use With (Application) | Storing, Packing, Taking Samples, Sampling and Transport of Pastes, Granules, Powders, Soil Samples and Frozen Goods (-40°C up to Briefly +95°C) |
Product Code | Brand | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
10096471
|
Buerkle™
6250-0500 |
500 mL | 103 x 103 x 64 mm |
3.75€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10475582
|
Buerkle™
6250-1000 |
1000 mL | 103 x 103 x 127 mm |
5.03€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10659012
|
Buerkle™
6250-1001 |
1000 mL | 208 x 103 x 64 mm |
7.15€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10310642
|
Buerkle™
6250-2000 |
2000 mL | 208 x 208 x 64 mm |
11.28€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10023373
|
Buerkle™
6250-2300 |
2000 mL | 255 x 103 x 94 mm |
10.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10390682
|
Buerkle™
6250-3300 |
3200 mL | 208 x 208 x 94 mm |
14.20€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10770583
|
Buerkle™
6250-1500 |
1500 mL | 208 x 103 x 94 mm |
9.55€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Frozen goods from -40° up to briefly +95°C
- Ideal use of space, can be stacked
- Comes complete with press-in lid
- Transparent in color
- Can be deep frozen
- Smooth surface, no crevices
Specifications
Press-in Lid | |
Polyethylene | |
Storing, Packing, Taking Samples, Sampling and Transport of Pastes, Granules, Powders, Soil Samples and Frozen Goods (-40°C up to Briefly +95°C) | |
All-purpose Boxes |
Transparent | |
Square | |
Press-in Lid |