Learn More
Buerkle™ LDPE Narrow-Necked Bottle
Product Code. 15435554 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
10 mL
20 mL
30 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Buerkle™ LDPE Narrow-Necked Bottle for storing, packing, lab, taking samples, sampling and transport of liquids.
  • Universal bottles for storing, packing, lab, taking samples, sampling and transport of liquids.
  • With screw cap
Specifications
Capacity (Metric) 100 mL
Color Transparent
Diameter (Metric) Outer Neck 18 mm
Diameter (Metric) Inner Neck 13 mm
For Use With (Application) Foodstuffs
Includes Screw cap
Height (Metric) With Closure 106 mm
Material LDPE
Neck ID (Metric) 13 mm
Replacement Closure 0300-1018
Quantity 100/Pk.
Weight (Metric) 0.0145 kg
Thread Size (Metric) 18 mm
Closure Color Transparent
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 106 mm
Closure Material Polyethylene
Diameter (Metric) 45 mm
Closure Type Screw Cap