Learn More
Kartell™ LDPE Bucket with Spout
Graduated bucket constructed of sturdy LDPE plastic. Kartell™ Buckets with Spout provide a durable choice for laboratory cleaning solutions.
12.55€ - 24.10€
Specifications
Product Type | Bucket |
---|---|
Graduations | Yes |
Material | LDPE |
Color | White |
Quantity | 1 Ea. |
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
10747152
|
Kartell™
0090500 |
9 L | 250 mm |
12.55€
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
||||
10132381
|
Kartell™
0090600 |
280 mm |
16.55€
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
10597151
|
Kartell™
0090700 |
310 mm |
24.10€
Each |
Estimated Shipment: 15-12-2025
Log in to see stock. |
|||||
Description
- Variety of sizes
- Suitable for foodstuff
- Integral deep spout for easy pour
Specifications
Bucket | |
LDPE | |
1 Ea. |
Yes | |
White |