Learn More
Logistik™ Lancer-Style Chemical Splash Laboratory Coats
Provide the answer to full frontal protection with Logistik™ Lancer-Style Chemical Splash Laboratory Coats The wrap-over style Lancer front coat helps repel liquid chemicals and provides the wearer with an excellent level of protection since the use of chemicals in a laboratory can prove a constant hazard to standard style open neck coats.
Brand: Logistik™ CS849WHM
Description
- Full frontal protection
- Chem-Tech material
- Fluorochemical liquid repellent
Specifications
General Purpose Lab Coat | |
Unisex | |
White | |
Reusable | |
Lancer | |
Open | |
101.6 to 104.14 cm | |
100 cm |
Medium | |
Polyester, Carbon | |
Stud-fastened | |
Non-sterile | |
2 | |
Fluorochemical Liquid Repellent | |
CE EN 13034 | |
65% Polyester, 35% Cotton |