Learn More
Brady™ Laminated Polyester Fire Safety Signs: Fire Blanket Pictogram
3.70€ - 5.20€
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
11700574
|
Brady™
250529 |
148L x 148 mmH |
4.00€
Each |
In Stock
Log in to see stock. |
|||||
10204604
|
Brady™
250530 |
200L x 200 mmH |
5.20€
Each |
In Stock
Log in to see stock. |
|||||
10783985
|
Brady™
250528 |
100L x 100 mmH |
3.70€
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||