Learn More
Thermo Scientific SMALL VOLUME BPC 3 PORTS 20L Injection Port , 20 L
Brand:  Thermo Scientific SH30709.03
SMALL VOLUME BPC 3 PORTS 20L Injection Port , 20 L
Additional Details : Weight : 0.45000kg
Product Code. 10399673