Learn More
Liebherr™ KTS149X Model ABS Refrigerator
Brand: Liebherr™ KTS 149
This item is not returnable.
View return policy
Specifications
General Purpose Refrigerator | |
3°C to 8°C | |
R 600a | |
ISO 9001 | |
KTS149X | |
850/820 mm | |
230 V | |
50 Hz |
128 L | |
Automatic | |
230V | |
White | |
623 mm | |
554 mm | |
230 V 50 Hz | |
40 kg |