Learn More
KleenGuard™ A20+ Coveralls
Provides general protection against powders, larger fibres, radioactive dust and chemical splashes. Kimberly-Clark™ Kleenguard™ A20+ Coveralls are perfect for general maintenance, cleaning and construction utility areas.
161.14€ - 166.24€
Specifications
Product Type | Single-use Coveralls |
---|---|
Gender | Unisex |
Disposable | Single-use |
Sterility | Non-sterile |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
13474556
|
KleenGuard™
95150 |
Small |
166.24€
Pack of 25 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
13460759
|
KleenGuard™
95200 |
3X-Large |
161.14€
Pack of 25 |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||||
Description
- Breathable, silicone-free coveralls, with an elasticated hood, ankle and wrists for comfort, safety and strength
- Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 5 and 6 by CEN
- Protects against powders, larger fibres, radioactive dust and chemical splashes
- Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
- Silicone-free, low lint and treated to be antistatic - to EN 1149-5
- Single-use protective clothing to improve safety and comfort in high-risk areas. Reduces contamination in critical areas
- Offers complete freedom of movement with comfortable body design
- Ideal for working in general manufacturing and maintenance
- Tripled-stitched seams help to protect against tearing with an easy-fastening snap front
Specifications
Single-use Coveralls | |
Unisex | |
Single-use | |
Non-sterile |