Learn More
DWK Life Sciences Kimble™ Kontes™ Glass Plug Gas Balloon Adapters
With #1 hose connection
275.00€
Specifications
Certifications/Compliance | USP Type I, ASTM E438, Type I, Class A |
---|---|
For Use With (Application) | For protective storage in jointed vessels |
Fits Tubing ID (English) | 0.25 in. |
Fits Tubing ID (Metric) | 6.35mm |
Includes | Hose connection 1, balloon |
Product Code | Brand | Standard Taper Joint No. | Height (English) | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Height (English) | Width (English) | Price | Quantity & Availability | |||
11842933
|
DWK Life Sciences
2112502440 |
24/40 | 5.98 in. | 2.67 in. |
275.00€
Each |
Estimated Shipment: 24-02-2026
Log in to see stock. |
|||
Description
Kimble-Chase Kontes™ Glass Plug Gas Balloon Adapter
Specifications
USP Type I, ASTM E438, Type I, Class A | |
0.25 in. | |
Hose connection 1, balloon | |
Inlet Adapter |
For protective storage in jointed vessels | |
6.35mm | |
Borosilicate Glass |