Learn More
DWK Life Sciences Kimble™ KIMAX™ Reusable Volumetric Unserialized Class A Pipets
Kimax-51 glass, with tempered tip and top-end contouring.
104.00 €
Specifications
Calibration | To Contain, To Deliver |
---|---|
Certifications/Compliance | ASTM E969 |
Class | A |
Material | Borosilicate Glass |
Tolerance | ±0.01 mL |
Product Code | Brand | Volume (Metric) | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
10067642
|
DWK Life Sciences
370073 |
3 mL | 13.77 in. | 35 cm | N/A | ||||
Description
To Contain and To Deliver; Chip-resistant tempered ends.
- Standard Class A volumetric pipets
- Made from chemical- and abrasion-resistant 51 expansion glass
- Calibrated “To Contain” (bottom line) and “To Deliver” (top line) according to Class A tolerances
- Color-coded graduations make it easy to sort and identify different sizes
- With tempered tip and top-end contouring for strength and chip-resistance
Specifications
To Contain, To Deliver | |
A | |
±0.01 mL | |
Reusable |
ASTM E969 | |
Borosilicate Glass | |
Unserialized Volumetric Pipet |