Learn More
BUCHI Rotavapor I-300 Vacuum Controller and Interface
The BUCHI Interface I-300 controls all distillation parameters of a Rotavapor R-300 system including the optional V-300 Vacuum Pump and F-3XX series chillers.
1330.32€ - 3094.64€
Product Code | Brand | Includes | For Use With (Equipment) | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | For Use With (Equipment) | Model | Price | Quantity & Availability | |||
15540600
|
BUCHI
11I300S10 |
Kit for V-300/600 | R-300, V-300, V-600 | Standard |
1996.10€
Each |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||
15550600
|
BUCHI
11I300S20 |
Kit for V-300/600, Woulff Bottle | R-300, V-300, V-600 | Standard |
2221.50€
Each |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||
15560600
|
BUCHI
11I300S30 |
Valve Unit; Use: Non-BUCHI Vacuum Source | R-300, Non-BUCHI Vacuum Pumps | Standard |
2694.72€
Each |
Estimated Shipment: 01-09-2025
Log in to see stock. |
|||
15590590
|
BUCHI
11I300P00 |
Standalone | R-300, V-300, V-600 | Pro |
1757.06€
Each |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||
15520600
|
BUCHI
11I300P30 |
Valve Unit | R-300 with Non-BUCHI Vacuum Pumps | Pro |
3094.64€
Each |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||
15530600
|
BUCHI
11I300S00 |
Standalone | R-300, V-300, V-600 | Standard |
1330.32€
Each |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||
15510600
|
BUCHI
11I300P20 |
Kit for V-300/600 Woulff Bottle | R-300 | Pro |
2631.07€
Each |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||
Description
- Easy plug and play integration into a Rotavapor R-300 system at any time
- Central control of all process parameters such as rotation speed, temperature (heating, cooling, vapor) and pressure
- Remote monitoring on smart phones including push notifications and live view
- Modes: manual, timer, continuous
- Flexibility of the interface allows easy installation and convenient operation from multiple positions:
- Rotavapor handle
- Vacuum pump
- Fume hood or lab bench wall
- Stand-alone operation to locally control a central vacuum system
- Synchronized processing thanks to centrally controlled rotary evaporator, heating bath, speed-controlled vacuum pump and recirculating chiller
- Unattended operation (optional):
- Automatic distillation with CloudDest sensor
- Foam sensor for foaming samples