Learn More
Mettler Toledo™ InLab™ Smart Pro-ISM pH Electrode
Brand: Mettler Toledo™ 30027775
1050.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
K8SD | |
InLab™ Smart Pro-ISM pH Electrode | |
0 to 14 | |
ISO 9001, CE certified |
12 mm | |
Glass | |
0°C to 130°C |