Learn More
Fisherbrand™ Individual Stainless Steel Leaf Calibration Weights, Class E2 with certificate
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Brand:  Fisherbrand™ FB67818
60.15 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15575099
  • 83.00€ / 1 unit
Only null left
Estimated Shipment: 24-09-2025
Add to basket
Description
Description
  • Class E2 calibration weight
  • In compliance with OIML R111
  • Delivered with a calibration certificate guaranteeing traceability
Specifications
Specifications
E2