Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2 with Certificate
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Brand: Fisherbrand™ FB67786
156.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Class E2 stainless steel calibration weight
- In compliance with OIML R111
- Delivered with a calibration certificate guaranteeing traceability
Specifications
E2 | |
200 g | |
Cylindrical |
Stainless Steel | |
200 g |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.