Learn More
OHAUS™ In Use Cover Guardian
Hotplate-Stirrer accessory
Brand: OHAUS™ 30500595
54.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
In Use Cover Guardian helps protect the control panel and allows for an easy clean
Specifications
Hotplate-Stirrer e-G51HS10C 120V US; Hotplate-Stirrer e-G71HS10C 120V US | |
Square, 10 x 10 | |
Cover | |
Metal |