Learn More
Brannan™ Hydrometer specific gravity shot loaded low adjustment for surface tension 0.950 to 1.000 x 0.001 S.G. 270mm Brannan
GREENER_CHOICE
Brand:  Brannan™ 10/923/0
17.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12801468
  • 17.85 EUR / Each
Only null left
Estimated Shipment: 08-10-2025
Description
Hydrometer specific gravity shot loaded low adjustment for surface tension 0.950 to 1.000 x 0.001 S.G. 270mm