Learn More
Hydrobromic acid, Reagent Grade, 48%, Honeywell™
Product Code. p-7111780 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
500mL
Reagent Grade, 48%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10035-10-6, 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br
Specifications
Boiling Point 126°C
CAS Min % ≥50.0000%, ≥30.0000%
CAS Max % <70.0000%, <50.0000%
Density 1.49 g/mL at 25 °C(lit.)
Packaging Glass Bottle
Quantity 500 mL
Linear Formula I2
UN Number UN1788
Beilstein 3587158
IUPAC Name hydrogen bromide
Formula Weight 80.91
Grade Reagent
Chemical Name or Material Hydrobromic acid, Reagent Grade, 48%