Learn More
Sperian™ Honeywell™ XC™ Clear Safety Glasses
Click to view available options
Frame Color:
Blue
Unit Size:
Each
Description
- Adjustable temples and lenses with flexible noise piece
- Extra flexible blue frame of plastic
- Easily replaceable lenses - can be changed without replacing the entire frame
- All pressure points are relieved for exceptional all-day comfort
Specifications
Specifications
Certifications/Compliance | EN 166 and EN 170 1F 3-1.2 |
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Product Type | Eye and Face Protection Spectacles |
For Use With (Application) | General use and protection against impact (45m/s), Mechanical work (grinding, turning, milling) |
Frame Color | Blue |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Frame Material | Plastic |