Learn More
Sperian™ Honeywell™ A700 Series Clear Hard Coat Safety Glasses
Click to view available options
Frame Color:
Clear
Unit Size:
Pack of 10
Description
- Affordably priced and economical for your business
- Lightweight design with sport temples and soft temple tip pads
- Moulded universal nose bridge
- Provides a secure, wraparound fit for all-day comfort
- 8-base polycarbonate abrasion-resistant lenses
- Provides a 180° field of distortion-free vision
Specifications
Specifications
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Product Type | Eye and Face Protection Spectacles |
For Use With (Application) | General use and protection against impact (45m/s), Well suited for Indoor applications such as light manufacturing. |
Frame Color | Clear |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |