Learn More
Cole-Parmer™ Heating Block for Electrothermal™ RS-250D-S Shaking Reaction Stations
Accessory heating blocks to keep your reactions running smoothly.
2955.00€
Specifications
Description | Heating Block for Reaction System |
---|---|
For Use With (Equipment) | RS-250D-S Shaking Reaction Stations |
Type | Heating Block |
Warranty | 1 Year |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11894770
|
Cole-Parmer™
99968-05 |
4 x 96-well PTFE Microplates |
2955.00€
Each |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
11791467
|
Cole-Parmer™
99968-07 |
96 x 16 mm Tubes |
2955.00€
Each |
Estimated Shipment: 10-10-2025
Log in to see stock. |
|||||
11889110
|
Cole-Parmer™
99968-08 |
40 x 24 mm Tubes |
2955.00€
Each |
Estimated Shipment: 14-11-2025
Log in to see stock. |
|||||
Description
- Compatible with Electrothermal RS9000 Reaction Stations
Specifications
Heating Block for Reaction System | |
RS-250D-S Shaking Reaction Stations | |
Heating Block | |
1 Year |