Learn More
BRAND™ HDPE Wide Neck Storage Bottles
HDPE construction. Wide neck bottle includes screw cap and carrying handle.
Brand: BRAND™ 130970
27.63 EUR valid until 2025-12-31
Use promo code "24323" to get your promotional price.
This item is not returnable.
View return policy
Description
- HDPE construction
- Wide neck
- Includes screw cap and carrying handle
Specifications
5 L | |
HDPE | |
Storage Bottle |
Carrying handle | |
Yes | |
Screw Cap |