Learn More
Masterflex™ HDPE T-Connector Barbed Fittings
Barbed T connectors are simple and economical fittings designed for quick and easy assembly of fluid paths in the laboratory.
Brand: Masterflex™ 30623-60
This item is not returnable.
View return policy
Description
- Use with low-pressure applications, generally less than 30 psi (2 bar)
- Manufactured from quality HDPE
- Sterilization by ethylene oxide (ETO) only
- Complete range of standard sizes available
- Provided in packs of 10
Specifications
HDPE | |
10 Pack | |
T-shape |
3/4 in. tubing I.D. | |
Ethylene Oxide | |
Barbed Fitting |