Learn More
Hamilton™ Disposable Tip Adapter
Designed to enable the connection of a standard 1mL disposable pipette tip to the end of a Hamilton™ syringe.
Brand: Hamilton™ 31330
29.52 EUR valid until 2025-09-21
Use promo code "24100" to get your promotional price.
Description
- Adapter connects to standard Luer Tip (LT) and PTFE Luer Lock (TLL) syringes
- Ideal for eliminating the possibility for cross-contamination
- Manufactured from CTFE
- Provided individually
Specifications
CTFE | |
Luer-tip Adapter | |
100 μL | |
PB600-1 Repeating Dispensers |