Learn More
Lenz Laborglasinstrumente™ Ground Joint Sleeve with Grip Collar
Sturdy design for permanent use, with grip collar and sealing rings for excellent tightness
Brand: Lenz Laborglasinstrumente™ 01101745
52.11 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Outstanding chemical resistance at operating temperatures from -200 to +260 °C
- With grip collar and sealing rings for excellent tightness
Specifications
For ground joints NS 45/40 | |
1 | |
45/40 |
PTFE | |
-200 to +260 °C |