Learn More
Kinematica™ Stainless Steel Mixing attachment for MB 800 Laboratory Mixer
Mixing attachment for MB 800 consists of stainless steel, gasket and blade head as well as lid with opening for material addition.
820.00€ - 1215.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11999058
|
Kinematica™
36050055 |
4000 mL |
1215.00€
Each |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||||
11989058
|
Kinematica™
36050050 |
2000 mL |
820.00€
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
Specifications
Stainless Steel | |
MB 800 |