Learn More
Graphite plate, pyrolytic, 1.27x9.98x9.98mm (0.05x0.393x.393in), Thermo Scientific Chemicals
Solid Plate for SEM calibration
255.27€ - 841.00€
Specifications
Chemical Name or Material | Graphite plate, pyrolytic |
---|---|
Name Note | 1.27x9.98x9.98mm (0.05x0.393x.393 in.) |
MDL Number | MFCD00144065 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15466315
|
Thermo Scientific Alfa Aesar
040156.KF |
1 Pc. |
255.27€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15476315
|
Thermo Scientific Alfa Aesar
040156.KJ |
5 Pc. | N/A | ||||||
Description
Graphite plate is used for fuel cells, electrolysis, electroplating, and other various mechanical applications where friction, heat, or electrical properties are factors. It is also used as a solid plate for SEM calibration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Graphite plate, pyrolytic | |
1.27x9.98x9.98mm (0.05x0.393x.393 in.) | |
MFCD00144065 | |
Insoluble in water. |
Safety and Handling
missing translation for 'einecsNumber' : 231-955-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures