Learn More
Gold wire, 14kt, red, 1.63mm (0.064 in.) dia., Au 58.3% min, Thermo Scientific™
Click to view available options
Quantity:
10 cm
25 cm
50 cm
Unit Size:
10cm
25cm
50cm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Chemical Name or Material | Gold wire (14kt) |
Color | Red |
Name Note | 1.63mm (0.064 in.) dia. |
MDL Number | MFCD00003436 |
Quantity | 10 cm |
Solubility Information | Insoluble in water. |
Concentration or Composition (by Analyte or Components) | % Concentration Cu>Ag, Au ≤58.3% |
Weight | ≈28g/m |
Form | Wire |