Learn More
Glycerol monoacetate, tech., mixture of isomers, cont. varying amounts of diacetate, Thermo Scientific Chemicals
Product Code. 11417906
Click to view available options
Quantity:
250 g
1000 g
CAS: 26446-35-5 | C5H10O4 | 134.13 g/mol
Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.
Solubility
Soluble in water, methanol, ethanol, ether (slightly), and insoluble in benzene.
Notes
Hygroscopic. Protect from moisture. Store at room temperature. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26446-35-5
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
MDL Number MFCD00036185
InChI Key KMZHZAAOEWVPSE-UHFFFAOYNA-N
Synonym monoacetin, acetin, 1-monoacetin, glycerol monoacetate, glyceryl monoacetate, acetoglyceride, glyceryl acetate, acetin, 1-mono, monacetin, glycerol 1-monoacetate
PubChem CID 33510
IUPAC Name 2,3-dihydroxypropyl acetate
SMILES CC(=O)OCC(O)CO
Specifications
Density 1.21
Boiling Point 258°C
Flash Point 145°C (293°F)
Refractive Index 1.45
Quantity 1000 g
Sensitivity Hygroscopic
Merck Index 14,6242
Solubility Information Soluble in water,methanol,ethanol,ether (slightly),and insoluble in benzene.
Formula Weight 134.13
Grade Technical
Assay cont. varying amounts of diacetate
Chemical Name or Material Glycerol monoacetate, tech., mixture of isomers