Learn More
Hecht Karl™ Glass Shot
36.00€
Specifications
Type Shot
Material Glass
Quantity 1 kg
View More Specs
Products 2
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
10368911
View Documents
Hecht Karl™
41401010
10 mm
36.00€
1kg
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 12-09-2025
Add to basket
10777322
View Documents
Hecht Karl™
41401008
8 mm
36.00€
1kg
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 04-09-2025
Specifications
Shot
Glass
1 kg