Learn More
Fisherbrand™ Glass Probe
For Use With: Digital Thermoregulator VTF and VTF EVO
Brand:  Fisherbrand™ A00000003
151.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11755674
  • 157.00€ / Each
Only null left
Estimated Shipment: 15-09-2025
Description
  • 250 mm long
  • Immersible in liquid
Specifications
Specifications
Digital Thermoregulator VTF and VTF EVO
250 mm
Probe