Learn More
Sartorius™ Glass Bell Jar
Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
100g to 1kg Weights
10kg Weights
1g to 50g Weights
1mg to 5g Weights
20kg Weights
2kg to 5kg Weights
50kg Weights
Unit Size:
Each
Specifications
Specifications
Product Type | Bell Jar |
Material | Glass |
Shape | Bell |
For Use With (Equipment) | 10kg Weights |
Quantity | 1 Each |