Learn More
BRAND™ Glass Automatic Burette - BB
Shop All Brand Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
Description
Specifications
Specifications
Capacity (Metric) | 25 mL |
Material | Glass |
Graduations | 0.05 mL |
Includes | Bottle |
Type | Automatic Buret |